missing translation for 'onlineSavingsMsg'
Learn More

WAY 100635 maleate, Tocris Bioscience™

Potent, 5-HT1A silent antagonist. Also D4 agonist

Brand:  RD Systems (Bio-Techne) 4380/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464654

  • £586.21 / 50mg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Potent, silent antagonist of 5-HT 1A receptors (IC 50 = 2.2 nM; K i = 0.84 nM for rat 5-HT 1A receptors). Displays 100-fold selectivity for 5-HT 1A over other 5-HT subtypes. Also exhibits agonist activity at dopamine D 4 receptors.
Specifications

Chemical Identifiers

634908-75-1
540.661
N-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinylcyclohexanecarboxamide maleate
formic acid;N-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-N-pyridin-2-ylcyclohexanecarboxamide;prop-2-enoic acid
C29H40N4O6
QYOBEWLYELTWTQ-UHFFFAOYSA-N
77518953
COC1=CC=CC=C1N2CCN(CC2)CCN(C3=CC=CC=N3)C(=O)C4CCCCC4.C=CC(=O)O.C(=O)O

Specifications

WAY 100635 maleate
634908-75-1
N-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinylcyclohexanecarboxamide maleate
COC1=CC=CC=C1N2CCN(CC2)CCN(C3=CC=CC=N3)C(=O)C4CCCCC4.C=CC(=O)O.C(=O)O
540.661
538.64
50mg
C29H40N4O6
QYOBEWLYELTWTQ-UHFFFAOYSA-N
formic acid;N-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-N-pyridin-2-ylcyclohexanecarboxamide;prop-2-enoic acid
77518953
>99%
Videos