missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow Single-Segment Manifold Tubing for Peristaltic Pumps 205S/CA and 205U/CA: PVC-Solvent Resistant

Brand:  Watson-Marlow 9840279000

Code : X3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12716017

  • £48.65 / Each


Estimated Shipment
06-04-2018
Add to basket

Specifications

Specifications

Single Segment Manifold Tubing
Purple/White
2.79mm
0.117 to 21mL/min.
205U/CA low flow multi channel pump
PVC
Videos