missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow Manifold Tubing for Pump Models 403U/VM2, 403U/VM3 and 403U/VM4

Brand:  Watson-Marlow 980.0254.000

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12496310

  • £76.85 / Pack of 12


Estimated Shipment
12-02-2018
Add to basket

Specifications

Specifications

PVC
Purple/Orange
2.54mm
Tubing
205S/CA low flow multi channel pump
Videos
Documentation