missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ultrapure, Spectrophotometric Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Description
Water ultrapure is used in spectrophotometry and environmental testing. It is also used in conductometric titration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Solubility Information | Miscible with methanol,acetone,ethanol. |
Formula Weight | 18.02 |
Grade | Spectrophotometric |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Density | 1 |
Quantity | 4 x 1 L |
Refractive Index | 1.333 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'