missing translation for 'onlineSavingsMsg'
Learn More

Water, ultrapure, Spectrophotometric Grade, Thermo Scientific Chemicals

Catalog Number 11333557
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Maximum level of impurities: Evaporation residue 2ppm, Specific conductance at 25° 1.5μ mhos/cm, SiO2 0.01ppm, Heavy metals (as Pb) 0.01ppm, Substances reducing permanganate P.T.

Water ultrapure is used in spectrophotometry and environmental testing. It is also used in conductometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Solubility Information Miscible with methanol,acetone,ethanol.
Formula Weight 18.02
Grade Spectrophotometric
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1
Quantity 4 L
Refractive Index 1.333
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'