missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, TraceSELECT™, For Trace Analysis, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
10 L
4 x 2.5 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle
Description
High purity solvents for speciation analysis by LC-ICP-MS
Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance
The blank values for metal traces in these solvents are in the ppb range or lower.
Specifications
pH | 6 to 8 (at 20°C) |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
UN Number | NONH for all modes of transport |
Beilstein | 2050024 |
Formula Weight | 18.02g/mol |
Grade | TraceSELECT™ |
Odor | Odorless |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'