missing translation for 'onlineSavingsMsg'
Learn More

Water, for qPCR or RT-qPCR, Fisher BioReagents

Catalog Number p-8357044
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly Bottle
Poly CUBE
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

High purity water rigorously tested for degrading enzymes and genomic DNA that interefere with amplification and can lead to false-positive signals in PCR, qPCR and RT-qPCR applications.

  • Minimize the risk of false positive signals
  • Tested for DNase, RNase, protease, nickase, and E. coli and human genomic DNA using sensitive assays
  • Evaluated for metal impurities at low ppb concentrations to ensure optimal enzymatic activity, especially for polymerase during PCR
  • Sub-micron filtered to remove particulates
  • Suitable for use in qPCR and RT-qPCR for genomics work, microarrays, and other sensitive life science applications
  • Also useful for the manufacturing of PCR-related kits and mastermixes
TRUSTED_SUSTAINABILITY
pH 5.4 to 7.0
Color Clear, Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity ≤2μS/cm
DNase Not detected
Endotoxin <0.1EU/mL
Formula Weight 18.02
Grade qPCR
Packaging Poly Bottle
Quantity 1 L
Physical Form Liquid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'