Water, Microbial Cell Culture Grade, Fisher BioReagents


Overview
Brand: Fisher Bioreagents

Manufacturer Part Number: BP2820-500

UNSPSC: 12161500

Code: 32

Additional Details:
Additional Details: CAS Number: 7732-18-5 Weight: 0.55000kg


Packaging Nalgene Poly Bottle
Quantity 500mL

Product Code. 11343892

Quantity Price
1 £ 18.0 / 500mL
EU Stock 35
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Water
Name Note 0.03μm filtered and autoclaved
CAS 7732-18-5
Purity Grade Microbial Cell Culture
Molecular Formula H2O
Formula Weight 18.01

Specification

Formula Weight 18.01
Boiling Point 100°C
Quantity 500mL
Copper (Cu) 10 ppb max.
Bacteria < 1 CFU/mL
Yeast and Mold < 1 CFU/mL
Aluminum (Al) 10ppb max.
Barium (Ba) 10ppb max.
Tin (Sn) 10ppb max.
Sodium (Na) 20ppb max.
Growth Promotion Pass Test
Additional Information Vapor Pressure: 17.5mmHg at 20°C
Chromium (Cr) 10 ppb max.
Health Hazard 3 Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Rinse with water. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . .

NFPA

Health:0
Flammability:0
Instability:0
Conductivity 2μS/cm Max.
Manganese (Mn) 10 ppb max.
ChemAlert Storage Symbol Gray
Nickel (Ni) 10 ppb max.
Vapor Pressure 17.5mmHg at 20°C
CAS 7732-18-5
Magnesium (Mg) 10 ppb max.
Purity Grade Microbial Cell Culture
Calcium (Ca) 20 ppb max.
Iron (Fe) 10 ppb max.
Potassium (K) 10 ppb max.
Resistivity 16 MEGOHM-cm min.
Selenium (Se) 10ppb max.
Arsenic (As) 10ppb max.
Physical Form Liquid
Packaging Nalgene Poly Bottle
Health Hazard 2 Low hazard for usual industrial or commercial handling.
Filtered Through 0.03 Micron filtered and autoclaved
Molybdenum (Mo) 10 ppb max.
Molecular Formula H2O
Name Note 0.03μm filtered and autoclaved
Density 1.000g/cm³
Vanadium (V) 10ppb max.
Silver (Ag) 10ppb max.
Cobalt (Co) 10 ppb max.
Color Clear Colorless
Chemical Name or Material Water
Cadmium (Cd) 10 ppb max.
Oxidizable Substances Pass Test
Endotoxin 0.025 EU/mL max.
Lead (Pb) 10 ppb max.
Documentation Safety and Handling
ChemAlertStorageSymbol : Gray

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Rinse with water. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . .

NFPA

Health:0
Flammability:0
Instability:0