missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Water Jet Filter Pump
Withdraw liquids/gases, or create a vacuum. Bochem™ Water Jet Filter Pump is easy to use and assemble/disassemble.
Brand: Bochem™ 12954
Additional Details : Weight : 0.11100kg
Description
- With non-return valve.
- Resistant plastic.
- Consumption of 8L/minute.
- Simple assembly/disassembly.
Specifications
Connection 1/2inch, 3/4inch, tube diameter 12mm |