missing translation for 'onlineSavingsMsg'
Learn More

Water Blank For ICP, ASTM Type I, SPEX CertiPrep™

Catalog Number p-4385044
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • DI Water Matrix Blank
  • 500 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Grade Assurance
Packaging Triple-leached LDPE bottle
Solvent or Matrix H2O
Quantity 500 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Water Blank

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'