Learn More
Abnova™ WARS (Human) ELISA Kit
For the Quantitative detection of Human WARS.
Brand:  Abnova™ KA4174.1Kit
Additional Details : Weight : 0.02000kg
Product Code. 16008585
  • £527.81 / 1 set
Estimated Shipment
19-06-2020
Add to basket
Description
Description
  • WARS (Human) ELISA Kit is a sandwich enzymelinked immunosorbent assay for the quantificaiton of human WARS.
Specifications
Specifications
Human
7453
Store at -20°C
1 Kit
tryptophanyl-tRNA synthetase
Kit
WARS