missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Warning Tags: Do Not Operate

Indicate who is responsible for placing a Lockout device and who is authorized to remove it or make changes to it.

Brand:  Brady™ 225407

13.02 GBP valid until 2024-09-30
Use promo code "21675" to get your promotional price.


Product Code. 15931021

  • £13.70 / Pack of 10
Estimated Shipment: 07-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The tags are manufactured from a flexible polyester, resistant to water, oil, and extreme temperatures
  • Their matt surface allows you to write on them easily with a ball point or felt-tip pen
Specifications

Specifications

Red on White
110 mm
50 mm
Polyester
Cable Ties
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers