missing translation for 'onlineSavingsMsg'
Learn More

W-9 hydrochloride, Tocris Bioscience™

Calmodulin antagonist

Brand:  Tocris (Bio-Techne) 037050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437593

  • £429.24 / 50mg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Calmodulin antagonist (inhibits calmodulin-activated PDE activity with an IC50 of 72 μM).
Specifications

Chemical Identifiers

69762-85-2
377.324
w-9 hydrochloride, n-6-aminohexyl-5-chloronaphthalene-2-sulfonamide hydrochloride, n-6-aminohexyl-5-chloro-2-naphthalenesulfonamide, 2-naphthalenesulfonamide,n-6-aminohexyl-5-chloro-, hydrochloride 1:1, n-6-aminohexyl-5-chloro-2-naphthalenesulfonamide hydrochloride, w-7 isomer hydrochloride, n-6-aminohexyl-5-chloronaphthalene-2-sulfonamide-hydrogen chloride 1/1
N-(6-aminohexyl)-5-chloronaphthalene-2-sulfonamide;hydrochloride
C16H22Cl2N2O2S
FGWDLQXTCSKXSK-UHFFFAOYSA-N
44118677
C1=CC2=C(C=CC(=C2)S(=O)(=O)NCCCCCCN)C(=C1)Cl.Cl

Specifications

50mg
69762-85-2
w-9 hydrochloride, n-6-aminohexyl-5-chloronaphthalene-2-sulfonamide hydrochloride, n-6-aminohexyl-5-chloro-2-naphthalenesulfonamide, 2-naphthalenesulfonamide,n-6-aminohexyl-5-chloro-, hydrochloride 1:1, n-6-aminohexyl-5-chloro-2-naphthalenesulfonamide hydrochloride, w-7 isomer hydrochloride, n-6-aminohexyl-5-chloronaphthalene-2-sulfonamide-hydrogen chloride 1/1
C1=CC2=C(C=CC(=C2)S(=O)(=O)NCCCCCCN)C(=C1)Cl.Cl
377.324
377.33
W-9 hydrochloride
C16H22Cl2N2O2S
FGWDLQXTCSKXSK-UHFFFAOYSA-N
N-(6-aminohexyl)-5-chloronaphthalene-2-sulfonamide;hydrochloride
44118677
>98%
Videos
Documents