Learn More
Tocris Bioscience™ W-84 dibromide
Brand:  Tocris Bioscience™ 053250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13437693
  • £492.72 / 50mg
Estimated Shipment
27-06-2019
Description
Description
Stabilizes cholinergic antagonist-receptor complexes by an allosteric effect. Increases the protective effect of atropine against organophosphate poisoning.
Specifications
Specifications
Hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide
C32H44Br2N4O4
50mg
21093-51-6
Observed MW: 708.53
Soluble to 10mM in DMSO