Learn More
W-5 hydrochloride, Tocris Bioscience™
Calmodulin antagonist
Brand:  Tocris (Bio-Techne) 036810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487583
  • £133.65 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Calmodulin antagonist (inhibits Ca2+-calmodulin dependent PDE with an IC50 of 240 μM).
Specifications
Chemical Identifiers
61714-25-8
342.882
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride
C16H23ClN2O2S
HOCSVIGHWPLMFC-UHFFFAOYSA-N
173829
Specifications
W-5 hydrochloride
>98%
10mg
HOCSVIGHWPLMFC-UHFFFAOYSA-N
N-(6-aminohexyl)naphthalene-1-sulfonamide;hydrochloride
173829
61714-25-8
C16H23ClN2O2S
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride
C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)NCCCCCCN.Cl
342.882
342.88
Videos