Learn More
VUF 5681 dihydrobromide, Tocris Bioscience™
Potent H3 receptor silent antagonist
Brand:  Tocris (Bio-Techne) 249310
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13431334
  • £151.41 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Potent histamine H3 receptor silent antagonist (pKi = 8.35).
Specifications
Chemical Identifiers
639089-06-8
355.118
vuf 5681 dihydrobromide, 4-3-1h-imidazol-4-yl propyl piperidine dihydrobromide, 4-3-1h-imidazol-5-yl propyl piperidine-hydrogen bromide 1/2, 4-3-3h-imidazol-4-yl propyl piperidine dihydrobromide
4-[3-(1H-imidazol-5-yl)propyl]piperidine;dihydrobromide
C11H21Br2N3
YFFVIGXPKWSZHW-UHFFFAOYSA-N
53440140
C1CNCCC1CCCC2=CN=CN2.Br.Br
Specifications
10mg
639089-06-8
vuf 5681 dihydrobromide, 4-3-1h-imidazol-4-yl propyl piperidine dihydrobromide, 4-3-1h-imidazol-5-yl propyl piperidine-hydrogen bromide 1/2, 4-3-3h-imidazol-4-yl propyl piperidine dihydrobromide
C1CNCCC1CCCC2=CN=CN2.Br.Br
355.118
355.11
VUF 5681 dihydrobromide
C11H21Br2N3
YFFVIGXPKWSZHW-UHFFFAOYSA-N
4-[3-(1H-imidazol-5-yl)propyl]piperidine;dihydrobromide
53440140
>98%