missing translation for 'onlineSavingsMsg'
Learn More

VUF 5574, Tocris Bioscience™

Potent, selective hA3 antagonist

Brand:  Tocris (Bio-Techne) 135950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449223

  • £601.23 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective, competitive antagonist for the human adenosine A3 receptor (Ki = 4 nM). Displays ≥ 2500-fold selectivity over A1 and A2A receptors.
Specifications

Chemical Identifiers

280570-45-8
371.4
chembl70880, 1-2-methoxyphenyl-3-2-pyridin-3-yl quinazolin-4-yl urea, lopac-v-5888, d01aai, 1-2-methoxyphenyl-3-2-pyridin-3-ylquinazolin-4-yl urea, 3-2-methoxyphenyl-1-2-pyridin-3-ylquinazolin-4-yl urea, 1-2-methoxy-phenyl-3-2-pyridin-3-yl-quinazolin-4-yl-urea, n-2-methoxyphenyl-n'-2-3-pyridinyl-4-quinazolinyl-urea, n-2-methoxyphenyl-n'-2-3-pyrindinyl-4-quinazolinyl-urea, 3-2-methoxyphenyl-1-2-pyridin-3-yl quinazolin-4-yl urea
1-(2-methoxyphenyl)-3-(2-pyridin-3-ylquinazolin-4-yl)urea
C21H17N5O2
YRAFEJSZTVWUMD-UHFFFAOYSA-N
4046493
COC1=CC=CC=C1NC(=O)NC2=NC(=NC3=CC=CC=C32)C4=CN=CC=C4

Specifications

50mg
280570-45-8
chembl70880, 1-2-methoxyphenyl-3-2-pyridin-3-yl quinazolin-4-yl urea, lopac-v-5888, d01aai, 1-2-methoxyphenyl-3-2-pyridin-3-ylquinazolin-4-yl urea, 3-2-methoxyphenyl-1-2-pyridin-3-ylquinazolin-4-yl urea, 1-2-methoxy-phenyl-3-2-pyridin-3-yl-quinazolin-4-yl-urea, n-2-methoxyphenyl-n'-2-3-pyridinyl-4-quinazolinyl-urea, n-2-methoxyphenyl-n'-2-3-pyrindinyl-4-quinazolinyl-urea, 3-2-methoxyphenyl-1-2-pyridin-3-yl quinazolin-4-yl urea
COC1=CC=CC=C1NC(=O)NC2=NC(=NC3=CC=CC=C32)C4=CN=CC=C4
371.4
371.39
VUF 5574
C21H17N5O2
YRAFEJSZTVWUMD-UHFFFAOYSA-N
1-(2-methoxyphenyl)-3-(2-pyridin-3-ylquinazolin-4-yl)urea
4046493
>98%
Videos
Documents