missing translation for 'onlineSavingsMsg'
Learn More

VU 0365114, Tocris Bioscience™

Positive allosteric modulator of M5 receptors

Brand:  RD Systems (Bio-Techne) 4404/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13454704

  • £757.05 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective positive allosteric modulator of M5 (EC50 values are 2.7 μM for human M5, and >30 μM for M1, M2, M3 and M4 receptors).
Specifications

Chemical Identifiers

1208222-39-2
397.353
1-biphenyl-4-ylmethyl-5-trifluoromethoxy indoline-2,3-dione, 1-1,1'-biphenyl-4-ylmethyl-5-trifluoromethoxy-1h-indole-2,3-dione, 1-1,1'-biphenyl-4-ylmethyl-5-trifluoromethoxy indole-2,3-dione
1-[(4-phenylphenyl)methyl]-5-(trifluoromethoxy)indole-2,3-dione
C22H14F3NO3
SPBGRXOPAXZSER-UHFFFAOYSA-N
45281794
C1=CC=C(C=C1)C2=CC=C(C=C2)CN3C4=C(C=C(C=C4)OC(F)(F)F)C(=O)C3=O

Specifications

50mg
1208222-39-2
1-biphenyl-4-ylmethyl-5-trifluoromethoxy indoline-2,3-dione, 1-1,1'-biphenyl-4-ylmethyl-5-trifluoromethoxy-1h-indole-2,3-dione, 1-1,1'-biphenyl-4-ylmethyl-5-trifluoromethoxy indole-2,3-dione
C1=CC=C(C=C1)C2=CC=C(C=C2)CN3C4=C(C=C(C=C4)OC(F)(F)F)C(=O)C3=O
397.353
397.35
VU 0365114
C22H14F3NO3
SPBGRXOPAXZSER-UHFFFAOYSA-N
1-[(4-phenylphenyl)methyl]-5-(trifluoromethoxy)indole-2,3-dione
45281794
>98%
Videos
Documents