Learn More
VU 0364770, Tocris Bioscience™
Positive allosteric modulator at mGlu4
Brand:  RD Systems (Bio-Techne) 4469/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13444804
  • £124.20 / 10mg
Estimated Shipment
28-06-2019
Description
Description
Positive allosteric modulator at mGlu4 receptors (EC50 = 290 nM in mGlu4-expressing HEK 293 cells). Reverses haloperidol-induced catalepsy in rats; prevents attentional deficit and forelimb asymmetry in a rodent model of Parkinson's disease. Exhibits affinity for MAO-B and MAO-A (Ki values are 0.72 μM and 8.5 μM respectively). Brain penetrant.
Specifications
Chemical Identifiers
61350-00-3
232.667
n-3-chlorophenyl picolinamide, n-3-chlorophenyl-2-pyridinecarboxamide, n-3-chlorophenyl pyridine-2-carboxamide, 2-pyridinecarboxamide, n-3-chlorophenyl, d0w9vn, n-3-chloro-phenyl-pyridine-2-carboxamide
N-(3-chlorophenyl)pyridine-2-carboxamide
C12H9ClN2O
SUYUTNCKIOLMAJ-UHFFFAOYSA-N
836002
C1=CC=NC(=C1)C(=O)NC2=CC(=CC=C2)Cl
Specifications
VU 0364770
61350-00-3
n-3-chlorophenyl picolinamide, n-3-chlorophenyl-2-pyridinecarboxamide, n-3-chlorophenyl pyridine-2-carboxamide, 2-pyridinecarboxamide, n-3-chlorophenyl, d0w9vn, n-3-chloro-phenyl-pyridine-2-carboxamide
C1=CC=NC(=C1)C(=O)NC2=CC(=CC=C2)Cl
232.667
232.67
10mg
C12H9ClN2O
SUYUTNCKIOLMAJ-UHFFFAOYSA-N
N-(3-chlorophenyl)pyridine-2-carboxamide
836002
>99%
Videos