Learn More
VU 0357121, Tocris Bioscience™
Positive allosteric modulator of mGlu5
Brand:  RD Systems (Bio-Techne) 4437/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13464754
  • £135.24 / 10mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Positive allosteric modulator of mGlu5 receptors (EC50 = 33 nM). Binds to a site distinct from that bound by MPEP (Cat. No. 1212).
Specifications
Chemical Identifiers
433967-28-3
305.325
4-butoxy-n-2,4-difluorophenyl benzamide, d02vff, cambridge id 6991768, 4-butoxy-n-2,4-difluorophenyl-benzamide, benzamide, 4-butoxy-n-2,4-difluorophenyl, vu hplc
4-butoxy-N-(2,4-difluorophenyl)benzamide
C17H17F2NO2
AHCYOTLTLQTPSU-UHFFFAOYSA-N
2296132
CCCCOC1=CC=C(C=C1)C(=O)NC2=C(C=C(C=C2)F)F
Specifications
10mg
433967-28-3
4-butoxy-n-2,4-difluorophenyl benzamide, d02vff, cambridge id 6991768, 4-butoxy-n-2,4-difluorophenyl-benzamide, benzamide, 4-butoxy-n-2,4-difluorophenyl, vu hplc
CCCCOC1=CC=C(C=C1)C(=O)NC2=C(C=C(C=C2)F)F
305.325
305.32
VU 0357121
C17H17F2NO2
AHCYOTLTLQTPSU-UHFFFAOYSA-N
4-butoxy-N-(2,4-difluorophenyl)benzamide
2296132
>99%