missing translation for 'onlineSavingsMsg'
Learn More

VU 0240551, Tocris Bioscience™

KCC2 inhibitor

Brand:  Tocris (Bio-Techne) 388810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13443764

  • £162.36 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 560 nM for K+ uptake assay in KCC2-overexpressing cells). Exhibits selectivity over the Na-K-2Cl cotransporter, NKCCl. Also inhibits hERG and L-type Ca2+ channels.
Specifications

Chemical Identifiers

893990-34-6
342.435
n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio-acetamide, n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-ylthio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanylacetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanyl acetamide, d0m2pp, cid, vu hplc, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide
N-(4-methyl-1,3-thiazol-2-yl)-2-(6-phenylpyridazin-3-yl)sulfanylacetamide
C16H14N4OS2
WJRWSLORVIHRNX-UHFFFAOYSA-N
7211972
CC1=CSC(=N1)NC(=O)CSC2=NN=C(C=C2)C3=CC=CC=C3

Specifications

VU 0240551
893990-34-6
n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio-acetamide, n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-ylthio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanylacetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanyl acetamide, d0m2pp, cid, vu hplc, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide
CC1=CSC(=N1)NC(=O)CSC2=NN=C(C=C2)C3=CC=CC=C3
342.435
342.43
10mg
C16H14N4OS2
WJRWSLORVIHRNX-UHFFFAOYSA-N
N-(4-methyl-1,3-thiazol-2-yl)-2-(6-phenylpyridazin-3-yl)sulfanylacetamide
7211972
>99%
Videos