missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0155069
Potent and selective PLD1 inhibitor
POA
Specifications
Purity | >98% |
---|---|
CAS | 1781834-89-6 |
Product Type | VU 0155069 |
Molecular Weight (g/mol) | 499.43 |
Inhibitors | VU 0155069 |
Description
Potent and selective phospholipase D1 (PLD1) inhibitor (IC50 values are 46 and 933 nM for PLD1 and PLD2 respectively). Inhibits migration of human and mouse breast cancer cell lines in transwell assays.Specifications
>98% | |
VU 0155069 | |
VU 0155069 |
1781834-89-6 | |
499.43 |
For Research Use Only