Learn More
Tocris Bioscience™ VU 0155069
Potent and selective PLD1 inhibitor
Brand:  Tocris Bioscience™ 357550
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463224
  • £658.56 / 50mg
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
Potent and selective phospholipase D1 (PLD1) inhibitor (IC50 values are 46 and 933 nM for PLD1 and PLD2 respectively). Inhibits migration of human and mouse breast cancer cell lines in transwell assays.
Specifications
Specifications
1781834-89-6
VU 0155069
499.43
VU 0155069
>98