Learn More
VU 0155041, Tocris Bioscience™
Potent, positive allosteric mGlu4 agonist
Brand:  Tocris (Bio-Techne) 324810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13462644
  • £52.65 / 10mg
Estimated Shipment
29-03-2019
Add to basket
Description
Description
Potent, positive allosteric modulator/allosteric agonist at mGlu 4 receptors (EC 50 = 798 and 693 nM at human and rat mGlu 4 receptors respectively). Active in in vivo models of Parkinson's disease following i.c.v. administration. Also available as sodium salt (Cat. No. 3311 ).
Specifications
Chemical Identifiers
1093757-42-6
316.178
2-3,5-dichlorophenyl carbamoyl cyclohexane-1-carboxylic acid, cambridge id 7307507, 2-3,5-dichloroanilino carbonyl cyclohexanecarboxylic acid, cis-2-3,5-dichlorphenylcarbamoyl cyclohexanecarboxylic acid
2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid
C14H15Cl2NO3
VSMUYYFJVFSVCA-UHFFFAOYSA-N
2933724
C1CCC(C(C1)C(=O)NC2=CC(=CC(=C2)Cl)Cl)C(=O)O
Specifications
VU 0155041
1093757-42-6
2-3,5-dichlorophenyl carbamoyl cyclohexane-1-carboxylic acid, cambridge id 7307507, 2-3,5-dichloroanilino carbonyl cyclohexanecarboxylic acid, cis-2-3,5-dichlorphenylcarbamoyl cyclohexanecarboxylic acid
C1CCC(C(C1)C(=O)NC2=CC(=CC(=C2)Cl)Cl)C(=O)O
316.178
316.18
10mg
C14H15Cl2NO3
VSMUYYFJVFSVCA-UHFFFAOYSA-N
2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid
2933724
>99%
Videos