missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | VSIG4 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human VSIG4 Arg20-Pro283 Accession # Q9Y279 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
VSIG4 Polyclonal specifically detects VSIG4 in Human samples. It is validated for Western Blot.Specifications
VSIG4 | |
Goat | |
Polyclonal | |
IgG | |
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot | |
RUO |
Mouse myeloma cell line NS0-derived recombinant human VSIG4 Arg20-Pro283 Accession # Q9Y279 | |
Primary | |
Human | |
Unconjugated | |
11326 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only