Learn More
VPS45 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555619
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LLNQWTYQAM VHELLGINNN RIDLSRVPGI SKDLREVVLS AENDEFYANN MYLNFAEIGS NIKNLMEDFQ KKKPKEQQKL ESIADMKAFV ENYPQFK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
VPS45 Polyclonal antibody specifically detects VPS45 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
VPS45 | |
Polyclonal | |
Unconjugated | |
VPS45 | |
AI462172; AW554165; H1; H1VPS45; HGNC:14579; hlVps45; h-VPS45; leucocyte vacuolar protein sorting 45; mVps45; RP11-458I7.2; rVps45; SCN5; vacuolar protein sorting 45 (yeast); vacuolar protein sorting 45 homolog; vacuolar protein sorting 45 homolog (S. cerevisiae); vacuolar protein sorting 45A; vacuolar protein sorting protein 45; vacuolar protein sorting-associated protein 45; vesicular transport protein rvps45; Vps45; VPS45A; VPS45B; VPS54A; VSP45; Vsp45a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRW7 | |
VPS45 | |
Recombinant protein corresponding to Human VPS45 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only