Learn More
VPS11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558629
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HQHCFESYSE SDADCPTCLP ENRKVMDMIR AQEQKRDLHD QFQHQLKCSN DSFSVIADYF GRGVFNKLTL LTDPPTARLT SSLEAGLQRD LLMHSR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
VPS11 Polyclonal antibody specifically detects VPS11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
VPS11 | |
Polyclonal | |
Unconjugated | |
VPS11 | |
1200011A11Rik; END1; HLD12; hVPS11; PEP5; PP3476; RING finger protein 108; RNF108; vacuolar protein sorting 11 (yeast); vacuolar protein sorting 11 homolog; vacuolar protein sorting 11 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 11 homolog; VPS11; VPS11 CORVET/HOPS core subunit; VPS11, CORVET/HOPS core subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H270 | |
VPS11 | |
Recombinant protein corresponding to Human VPS11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only