missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Von Willebrand Factor Antibody (Ajinomoto patent anti-vWF) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28401-50ug
This item is not returnable.
View return policy
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Von Willebrand Factor | |
| Monoclonal | |
| Unconjugated | |
| P04275 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Ajinomoto patent anti-vWF | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| vWF | |
| 50 μg | |
| Cancer | |
| 7450 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction