Learn More
Poulten & Graf GmbH Volac™ Polystop™ Clear Dropping Bottles
Manufactured from soda lime glass. Poulten Graf™ Volac™ Polystop™ Dropping Bottles are available in two color varieties, a range of capacities and include an interchangeable 3-piece plastic stopper.
Brand: Poulten & Graf GmbH L188/B/10
Code : 02
Additional Details : Weight : 0.01000kg
Description
- Stopper consists of a strong ungraduated glass pipette, a durable vinyl teat and a PP-stopper
- Available in clear or amber glass in a variety of sizes
- Narrow neck construction
Specifications
Dropping Bottle | |
50 mL | |
Glass |
Stopper | |
Clear | |
16/16 |