Learn More
Poulten Graf™ Volac™ Polystop™ Clear Dropping Bottles
Manufactured from soda lime glass. Poulten Graf™ Volac™ Polystop™ Dropping Bottles are available in two color varieties, a range of capacities and include an interchangeable 3-piece plastic stopper.
Brand:  Poulten Graf™ L188/C/10
Code : 02
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12318208
  • £14.25 / Each
In Stock 280
Add to basket
Description
Description
  • Stopper consists of a strong ungraduated glass pipette, a durable vinyl teat and a PP-stopper
  • Available in clear or amber glass in a variety of sizes
  • Narrow neck construction
Specifications
Specifications
100mL
16/16
Stopper
Clear
Bottle, Dropping
Videos
Documentation