Learn More
Poulten Graf™ Volac™ Polystop™ Amber Dropping Bottles
Manufactured from soda lime glass. Poulten Graf™ Volac™ Polystop™ Dropping Bottles are available in two color varieties, a range of capacities and include an interchangeable 3-piece plastic stopper.
Brand:  Poulten Graf™ L188/AM/B/10
Code : 02
Additional Details : Weight : 0.06930kg
Product Code. 12338208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Stopper consists of a strong ungraduated glass pipette, a durable vinyl teat and a PP-stopper
  • Available in clear or amber glass in a variety of sizes
  • Narrow neck construction