Learn More
Poulten Graf™ Volac™ Glass Serological Pipettes
Offering economical single-use convenience leading to greatly reduced possibility of cross-contamination. Poulten Graf™ Volac™ Glass Serological Pipettes are disposable and have clear advantages of plastic counterparts in areas of chemical and temperature resistance, accuracy, transparency and clarity of markings.
Brand: Poulten Graf™ D807/PS
Code : 02
Additional Details : Weight : 0.01000kg
Description
- Ideal for use with radioactive and other hazardous materials
- Volumes are color-coded
- Environmentally friendly compared with plastic counterparts
- Graduated
Specifications
Glass | |
Yes | |
Cotton plug | |
5 mL | |
Single-use |
Sterile | |
0.05 mL | |
500 Pack | |
Serological Pipette |