missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Volac™ Class B Borosilicate Glass Burettes With Removable Stopcock

Brand:  Poulten Graf™ R9509/C

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12328378

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Class B
Removable PTFE-stopcock, PTFE key, Glass tip
PTFE
Burette
0.10mL
Borosilicate Glass
50mL
Videos
Documentation

Technical Documents