missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Volac™ Class A Soda Lime Glass Bulb Pipette without Works Certificate

Individually marked with the time of flow to transfer. Poulten Graf™ Volac™ Class A Soda Lime Glass Bulb Pipettes without Works Certificate are guaranteed to give both volumes and flow times within Class A limits.

Brand:  Poulten Graf™ R371/NC/K/2

Code : 01

Additional Details : Weight : 0.01000kg

Product Code. 12303478

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Tufftip treated to reduce chipping and breakage
  • Individually calibrated, tested and numbered for traceability
  • Color coded to BS 3996
Specifications

Specifications

BS1583, ISO648/Certificate of calibration
Yellow
2Pack
Bulb Pipette
Soda Lime Glass
A
Yes
±0.080mL
100mL
Videos
Documentation