missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Volac™ Class A Mixing Cylinders with Certificate

Brand:  Poulten Graf™ R264/WAC/I/2

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12308955

  • £158.84 / Pack of 2


Estimated Shipment
11-06-2018
Add to basket

Specifications

Specifications

Borosilicate GlassĀ 
A
Size Polypropylene Stopper, Batch Certificate, with Single Certificate
Polypropylene
ISO4788 - BS 604 - DIN 12680
1000mL
White Ceramic Glazed Ink Scale
10mL
Videos