missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Volac™ Class A Mixing Cylinders with Certificate

Brand:  Poulten Graf™ R264/WAC/F/2

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12378945

  • £52.69 / Pack of 2


Estimated Shipment
14-06-2018
Add to basket

Specifications

Specifications

Borosilicate GlassĀ 
A
29/32 Size Polypropylene Stopper
Polypropylene
ISO4788 - BS 604 - DIN 12680
250mL
White Ceramic Glazed Ink Scale
2mL
Videos