missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Volac™ Class A Borosilicate Glass Graduated Cylinders with Certificate

Brand:  Poulten Graf™ R263/WAC/I/2

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12398905

  • £242.00 / Pack of 2


Estimated Shipment
23-02-2018
Add to basket

Specifications

Specifications

Borosilicate GlassĀ 
A
Stopper, Works Certificate
Polypropylene
ISO4788 - BS 604 - DIN 12680
1000mL
White Ceramic Glazed Ink Scale
10mL
Videos