Learn More
Tocris Bioscience™ Voglibose
Orally active α-glucosidase inhibitor
Brand:  Tocris Bioscience™ 359650
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433264
  • £482.16 / 50mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Orally active α-glucosidase inhibitor (IC50 values are 3.9 and 6.4 nM at sucrase and maltase respectively). Increases glucagon-like peptide 1 (GLP-1) secretion and decreases food consumption in ob/ob mice, and reduces plasma concentrations of glucose, triglycerides and insulin in Wistar fatty rats. Exhibits antidiabetic and antiobesity activity in vivo.
Specifications
Specifications
Voglibose
Voglibose
267.28