missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VN1R4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555483
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TGKWNYTNIT VNEDLGYCSG GGNNKIAQTL RAMLL Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.
VN1R4 Polyclonal antibody specifically detects VN1R4 in Human samples. It is validated for Immunohistochemistry.Specifications
VN1R4 | |
Polyclonal | |
Unconjugated | |
VN1R4 | |
CTD-2245F17.9; G-protein coupled receptor GPCR27; hGPCR27; pheromone receptor; V1RL4; V1R-like 4; V1r-like receptor 4; VN1R4; vomeronasal 1 receptor 4; vomeronasal type-1 receptor 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
317703 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5H5 | |
VN1R4 | |
Recombinant protein corresponding to Human VN1R4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only