Learn More
VMP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556555
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LYLGPHIASV TLAAYECNSV NFPEPPYPDQ IICPDEEGTE GTISLWSIIS KVRIEACMWG IGTAIGELPP YFMARAARLS GAEPDDEEYQ EFEEMLEHAE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
VMP1 Polyclonal antibody specifically detects VMP1 in Human samples. It is validated for Immunohistochemistry.Specifications
VMP1 | |
Polyclonal | |
Unconjugated | |
VMP1 | |
3110098I04Rik; 4930579A11Rik; AI787464; EPG3; HSPC292; MIR21; mir-21a; NF-E2 induceble protein-2; NF-E2-inducible protein 2; ni-2; Protein ni-2; Tango5; TDC1; Tmem49; transmembrane protein 49; Vacuole membrane protein 1; VMP1; wu:fb05d08; wu:fb92g08; zgc:77153 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81671 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96GC9 | |
VMP1 | |
Recombinant protein corresponding to Human VMP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only