missing translation for 'onlineSavingsMsg'
Learn More

Mouse VLDLR Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication

POA

Specifications

Antigen VLDLR
Clone 268724
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Description

Description

VLDLR Monoclonal specifically detects VLDLR in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

VLDLR
LYOPH
Western Blot
Unconjugated
Rat
Mouse
P98156
7436
Mouse myeloma cell line NS0-derived recombinant mouse VLDL R Thr25-Ala798 Accession # P98156
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse VLDL R in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human ApoER2.
268724
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL R, VLDL receptor, VLDL-R, VLDLRCH
VLDLR
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only