Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
Brand: Sartorius™ VN01H41ETO
Code : NEW
Additional Details : Weight : 0.10000kg
Specifications
PCR | |
100 kDa | |
Polycarbonate, Polypropylene | |
DNA | |
0.1 to 0.5 mL |
Polycarbonate, Polypropylene | |
Concentration, Desalting, buffer exchange | |
25/Pk. | |
Vivacon 500 Centrifugal Unit |