missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 500, Sample Pack H (High MWCO)

Vivacon™ 500 are centrifugal units with Hydrosart™ membranes for fast DNA concentration and buffer exchange of samples of 100–500μL. Sample Pack H is intended for use with samples with a high molecular weight cut off, and include 4 units each of the 30, 50 and 100 kDa sets.

Brand:  Sartorius VN01HH12

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 11775046

  • £64.10 / 1 set

Estimated Shipment: 17-08-2022
to see stock.
Add to basket

Description

Description

Vivacon™ 500 is optimized for the concentration / recovery and buffer exchange applications of genomic DNA and peptides for samples up to 500μl initial volume. They are also the product of choice for peptide fractionation applications in this volume range. For optimal performance with highly diluted samples, e.g. forensic specimens, Vivacon™ 500 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux.

High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon™ 500 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as peptide fractionation (Filter Aides Sample Preparation, FASP1). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Concentration of up to 0.5 ml genomic DNA or peptide samples
  • Low binding material
  • Reverse spin possibility
  • MWCO printed on device
  • Complete and highly reproducible concentrate recovery
Specifications

Specifications

None
Polycarbonate Concentrator Body/Sleeve, Polypropylene Filtrate Vessel
30 to 100 kDa
Centrifugal UltraFiltration
Ultrafiltration Devices
5 μL
Hydrosart
0.1–0.5mL
DNA, Peptide
0.1 to 0.5 mL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers