missing translation for 'onlineSavingsMsg'
Learn More

VistaLab Technologies™ Reagent Reservoirs

Unique trough-within-trough design maximizes reagent recovery

Brand:  VistaLab Technologies™ 3054-1000

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15288304

  • £76.75 / Each


Estimated Shipment
21-02-2018
Add to basket

Specifications

Specifications

Yes
Polystyrene
Bulk
Yes
10/bag x 10; 100/Cs.
100mL
Pyrogen-free
3.38 oz., 100mL
Reagent Reservoir
Videos