missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs: 12-Channel

Product Code. 16425094
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually Wrapped
Quantity:
10/Cs.
25/Cs.
Unit Size:
Pack of 10
Pack of 25
This item is not returnable. View return policy

Product Code. 16425094

Brand: VistaLab™ Technologies 30541010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Unique trough-within-trough design maximizes reagent recovery.

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • 100mL model (Cat. No. 21-382-111) supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
  • 5 mL capacity / well

Specifications

Certifications/Compliance RNase/DNase, Pyrogen-free
Color Clear
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Height (English) 4 in.
Includes Lid
Length (English) 11 in.
Packaging Individually Wrapped
Quantity 10/Cs.
Volume (Metric) Well 5 mL
Width (English) 6 in.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.