VistaLab Technologies™ Pipet Tips for Adjustable- and Fixed-Volume Pipets
For use with Ovation™ adjustable (100-1000μL), fixed-volume (201-1000μL), and MLA™ Brand (201-1000μL) pipets
Overview
Brand: VistaLab Technologies™
Manufacturer Part Number: 40581000
UNSPSC: 41121600
Code: 04
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11996905
Quantity Price
1 £ 46.76 / Pack of 1000
Estimated Shipment
15-06-2017
Add to basket
Description and Specification
Specification
Volume (Metric) 35μL
Tip Style Micro
Packaging Bulk Pack, 1000 tips/bag
For Use With (Equipment) Ovation adjustable volume (0.2 to 2uL, 1.0 to 10uL, and 2.0 to 20uL) and fixed volume (1 to 20uL)