missing translation for 'onlineSavingsMsg'
Learn More

VistaLab Technologies™ Ovation™ Fixed-Volume Pipetter; Single Stroke

Naturally follows the contours of the palm

Brand:  VistaLab Technologies™ 11570010

Code : 04

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 11922225

  • £288.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Single-stroke operation.

  • Adjustable hook can be easily positioned for a custom fit for left- or right-handed users
  • Fits comfortably in the palm of your hand
  • Energy release button stores the energy captured during the acquisition of tips and uses energy to push off tip at the gentle push of a button
  • Significantly improves postures and forces associated with pipetting
  • Low-force, form-fitting plunger allows user to manually control the speed of aspirating and dispensing with minimal effort
  • Nozzle with replaceable filter prevents contamination of pipetter
  • Stands securely on the counter before, during, and after the job — no risk of contamination
  • In-lab calibration adjusted with internal software

Specifications

Specifications

±1.5%
Micro
Orange
0.50%
1 Year
10μL
Videos
Documentation