VistaLab Technologies™ Ovation™ Electronic Single Channel Pipette

Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions

Overview
Brand: VistaLab Technologies™

Manufacturer Part Number: 10651250L

UNSPSC: 41121507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11902225

Quantity Price
1 £ 708.55 / Each
Estimated Shipment
20-06-2017
Add to basket


Description and Specification

Specification

Accuracy ±0.75%
Display LCD
Volume (Metric) 25 to 1250μL
Model Left hand
Number of Channels 1
Battery Type Lithium Ion
Color Purple
Number of Speeds 5
Warranty 1 Year
Compatible Tips VistaLab™ tip
For Use With (Equipment) VistaLab™ tip
Increments 5μL
Finger Style Ergonomic
Documentation