missing translation for 'onlineSavingsMsg'
Learn More

VistaLab Technologies™ Ovation™ Electronic Single Channel Pipette

Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions

Brand:  VistaLab Technologies™ 1065-10MLL

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11982215

  • £1275.44 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • One-way disposable nozzle
  • Proprietary energy-release system discards tips at the push of a button
  • Contoured plunger electronically activates pipetting activity
  • Adjustable hook can be easily positioned for a custom fit and reduced holding effort
  • Bionatural shape gives user maximum stability when pipetting, with minimal contact pressure
  • Easy push-button controls
  • Fully electronic operations maximize efficiency for all tasks, including multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting, and automatic sequential dispensing
  • Stands securely on countertops

These electronic single channel Ovation models require a power supply unit 22-147-115 (sold separately) to recharge the pipettes battery.

Electronic single channel models require an Ovation Power Supply unit (Mfr. No. 9060-9001; sold separately) to recharge the pipetter's battery

Specifications

Specifications

±0.6%
VistaLab™ tip
Ergonomic
10μL
1 Year
Left handed
0.2 to 10mL
Lithium-ion
LCD
VistaLab™ tip
5
Red
1
Videos
Documentation