VistaLab Technologies™ Ovation™ Electronic Single Channel Pipette

Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions

Overview
Brand: VistaLab Technologies™

Manufacturer Part Number: 10650020L

UNSPSC: 41121507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11902215

Quantity Price
1 £ 708.55 / Each
Estimated Shipment
20-04-2017
Add to basket


Description and Specification

Specification

Battery Type Lithium Ion
Number of Speeds 5
Compatible Tips VistaLab™ tip
Color Yellow
Display LCD
Accuracy ±1%
For Use With (Equipment) VistaLab™ tip
Number of Channels 1
Model Left hand
Warranty 1 Year
Finger Style Ergonomic
Volume (Metric) 0.5 to 20μL
Speeds 5
Increments 0.05μL
Compatibility VistaLab™ tip
Number of Channels 1
Color Yellow
Documentation